Anti-APBB3 Rabbit Polyclonal Antibody
Catalog # ABGEAP20461C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:amyloid beta (A4) precursor protein-binding, family B, member 3
- Antigen symbol:APBB3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:internal
- Gene ID:10307
- Antigen synonyms:SRA|FE65L2
- Amino acid number:187 to 214
- Storage buffer:pH 7,4
- Molecular weight:52.585 kDa
- Sequence:GVGSSKGRDRDFAC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.44
- Shipping temperature:4 °C
- Immunogen:This APBB3 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 187-214 amino acids from the Central region of human APBB3.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: APBB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human