Order Entry
Export
ContactUsLinkComponent
Anti-MED26 Rabbit Polyclonal Antibody
Anti-MED26 Rabbit Polyclonal Antibody
Catalog # ABGEAP16422C
Supplier:  Abgent
Anti-MED26 Rabbit Polyclonal Antibody
Catalog # ABGEAP16422C
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Mediator Complex Subunit 26
  • Antigen symbol:
    MED26
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    internal
  • Gene ID:
    9441
  • Antigen synonyms:
    CRSP7|CRSP70
  • Amino acid number:
    191 to 220
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    65.446 kDa
  • Sequence:
    GSRLERDENDKHSGKC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.5
  • Shipping temperature:
    4 °C
  • Immunogen:
    This MED26 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 191-220 amino acids from the Central region of human MED26.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: MED26
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human