Anti-XYLB Rabbit Polyclonal Antibody
Catalog # ABGEAP17718A
Supplier: Abgent
Jump to:
Product Details & Documents
Type: Primary
Antigen: XYLB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Documents
Specifications
- Western blot:Yes
- Clonality:Polyclonal
- Flow cytometry:Yes
- Purification:Protein A & Peptide affinity purification
- Antibody type:Primary
- Epitope:N-terminal
- Amino acid number:18 to 47
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Host:Rabbit
- Sequence:YEESVHFDRDLPEFGC
- Concentration:0.35
- Molecular weight:58.382 kDa
- Shipping temperature:4 °C
- Antigen name:xylulokinase homolog (H. influenzae)
- Cat. No.:ABGEAP17718A
- Reactivity:Human
- Gene ID:9942
- Immunogen:This XYLB antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 18-47 amino acids from the N-terminal region of human XYLB.
- Conjugation:Unconjugated
- Antigen symbol:XYLB
- Storage buffer:pH 7,4
- ELISA:Yes



