Anti-TRIX3 Rabbit Polyclonal Antibody
ABGEAP19093A
: Abgent
- Antibody type:Primary
- Antigen symbol:TRIX3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Amino acid number:41 to 67
- Storage buffer:pH 7,4
- Sequence:KKTIQQRNLKTDC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.7
- Shipping temperature:4 °C
- Immunogen:This TRIX3 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 41-67 amino acids from the N-terminal region of human TRIX3.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Type: Primary
Antigen: TRIX3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human