Anti-Vtg Mouse Monoclonal Antibody [clone: ND-2D3]
Type: Primary
Antigen: Vtg
Clonality: Monoclonal
Clone: ND-2D3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: common Carp, fathead Minnow, goldfish, mullet, pinfish, sucker, Zebrafish
- Catalog No:
- CAYM170115-100
- CAYM170115-500
- Antigen symbol:
- VTG
- VTG
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- ND-2D3
- ND-2D3
- Reactivity:
- Zebrafish, Mullet, Carp, Sucker, Fathead minnow, Goldfish, Pinfish
- Zebrafish, Mullet, Carp, Sucker, Fathead minnow, Goldfish, Pinfish
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Vtg from plasma of 17-beta-estradiol treated striped bass (Morone saxatilis), brown bullhead (I. nebulosus), and common carp (C. carpio)
- Vtg from plasma of 17-beta-estradiol treated striped bass (Morone saxatilis), brown bullhead (I. nebulosus), and common carp (C. carpio)
- Purification:
- purified
- purified
- Cross adsorption:
- No
- No
- Storage buffer:
- 0,1 M sodium phosphate, pH 7,4, containing 0,15 M sodium chloride, 12.3 mM sodium azide, and 1% BSA
- 0,1 M sodium phosphate, pH 7,4, containing 0,15 M sodium chloride, 12.3 mM sodium azide, and 1% BSA
- Storage temperature:
- +4 °C
- +4 °C
- Shipping temperature:
- 4 °C
- 4 °C