Order Entry
Export
ContactUsLinkComponent
Anti-5-LO Rabbit Polyclonal Antibody
Anti-5-LO Rabbit Polyclonal Antibody
Catalog # CAYM160402-1
Supplier:  Cayman Chemical
Anti-5-LO Rabbit Polyclonal Antibody
Catalog # CAYM160402-1
Supplier:  Cayman Chemical

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    5-Methoxytryptophan
  • Antigen symbol:
    5-LO
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Hamster,
    Pig,
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Amino acid number:
    130 to 149
  • Sequence:
    QHRRKELETRQKQYRWMEWN
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    human and rat amino acids 130-149; The sequence is 95% homologous to 5-LO from mouse and hamster
  • Purification:
    purified by affinity chromatography
  • Size:
    1 Ea
  • Pk:
    1

Specifications

About this item

Type: Primary
Antigen: 5-LO
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Hamster, Human, Mouse, Pig, Rat