Order Entry
Export
ContactUsLinkComponent
Anti-COX-1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: CX111]
Anti-COX-1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: CX111]
  CAYM160111-250
Anti-COX-1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: CX111]
  CAYM160111-250

 

  • Antibody type:
    Primary
  • Antigen symbol:
    COX-1
  • Clonality:
    Monoclonal
  • Clone:
    CX111
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2b
  • Reactivity:
    Sheep,
    Human,
    Bovine,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Storage buffer:
    PBS, pH 7,2, with 50% glycerol, 0,5 mg/ml BSA and 0,02% NaN3
  • Storage temperature:
    +4 °C
  • Shipping temperature:
    4 °C
  • Immunogen:
    ovine COX-1
  • Size:
    250 µg
  • Pk:
    250 µG

 

 

Type: Primary
Antigen: COX-1
Clonality: Monoclonal
Clone: CX111
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Bovine, Human, Mouse, Sheep, Rat