Order Entry
Export
ContactUsLinkComponent
Anti-HCN4 Mouse Monoclonal Antibody [clone: S114-10]
Anti-HCN4 Mouse Monoclonal Antibody [clone: S114-10]
Catalog # CAYM13709-100
Supplier:  Cayman Chemical
Anti-HCN4 Mouse Monoclonal Antibody [clone: S114-10]
Catalog # CAYM13709-100
Supplier:  Cayman Chemical

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    HCN4
  • Clonality:
    Monoclonal
  • Clone:
    S114-10
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat
  • Epitope:
    C-terminal
  • Cross adsorption:
    No
  • Amino acid number:
    1019 to 1108
  • Storage buffer:
    PBS, pH 7,4, containing 50% glycerol and 0,09% sodium azide
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    fusion protein amino acids 1,019-1,108 (cytoplasmic C-terminus) of rat HCN4
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: HCN4
Clonality: Monoclonal
Clone: S114-10
Conjugation: unconjugated
Epitope: C-terminal
Host: Mouse
Isotype: IgG
Reactivity: Human, Rat