Order Entry
Export
ContactUsLinkComponent
Anti-SIRT7 Rabbit Polyclonal Antibody
Anti-SIRT7 Rabbit Polyclonal Antibody
Catalog # CAYM13477-1
Supplier:  Cayman Chemical
Anti-SIRT7 Rabbit Polyclonal Antibody
Catalog # CAYM13477-1
Supplier:  Cayman Chemical

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Sirtuin 7
  • Antigen symbol:
    SIRT7
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Amino acid number:
    35 to 51, 361 to 377
  • Storage buffer:
    PBS containing 0,2% gelatin and 0,05% sodium azide
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    human SIRT7 amino acids 35-51 and 361-377
  • Purification:
    purified by Protein G chromatography
  • Size:
    1 Ea
  • Pk:
    1

Specifications

About this item

Type: Primary
Antigen: SIRT7
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human