Anti-MAGL Rabbit Polyclonal Antibody
Catalog # CAYM10212-500
Supplier: Cayman Chemical
Specifications
- Antibody type:Primary
- Antigen symbol:MAGL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:No
- Amino acid number:579 to 592
- Storage buffer:TBS, pH 7,4, containing 50% glycerol, 0,5 mg/mL BSA, and 0,02% sodium azide
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:human Nrf2 C-terminal amino acids 579-592
- Purification:purified
- Size:500 µl
- Pk:500 µl
Specifications
About this item
Type: Primary
Antigen: MAGL
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat