Anti-cPGES Rabbit Polyclonal Antibody
CAYM10209-500
- Antibody type:Primary
- Antigen symbol:cPGES
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:recombinant human MAGL
- Purification:purified by affinity chromatography
- Size:500 µl
- Pk:500 µl
Type: Primary
Antigen: cPGES
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope: Full length
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse