Anti-GST Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # CAYM10197-500
Supplier: Cayman Chemical
Specifications
- Antibody type:Primary
- Antigen name:Glutathione-S-Transferase
- Antigen symbol:GST
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:+4 °C
- Shipping temperature:4 °C
- Immunogen:glutathione S-transferase (S. japonicum)
- Size:500 µl
- Pk:500 µl
Specifications
About this item
Type: Primary
Antigen: GST
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: