Anti-EP4 Rabbit Polyclonal Antibody
CAYM101770-1
- Antibody type:Primary
- Antigen symbol:EP4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Sheep,Human
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:No
- Format:Serum
- Form:lyophilized
- Amino acid number:1 to 22
- Sequence:MSTPGVNSSASLSPDRLNSPVTI
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:human EP4 receptor N-terminal amino acids 1-22
- Size:1 Ea
- Pk:1
Type: Primary
Antigen: EP4
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human, Sheep