Anti-LAMP1 Mouse Monoclonal Antibody [clone: Ly1C6]
Type: Primary
Antigen: LAMP1
Clonality: Monoclonal
Clone: Ly1C6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Hamster, Rat
- Catalog No:
- CAYM10011443-25
- CAYM10011443-100
- Antigen symbol:
- LAMP1
- LAMP1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- Ly1C6
- Ly1C6
- Reactivity:
- Hamster, Rat
- Hamster, Rat
- Host:
- Mouse
- Mouse
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Immunogen:
- rat liver lysosomal membrane preparations
- rat liver lysosomal membrane preparations
- Cross adsorption:
- No
- No
- Storage buffer:
- PBS buffer, containing 0,09% sodium azide and 50% glycerol
- PBS buffer, containing 0,09% sodium azide and 50% glycerol
- Storage temperature:
- –20 °C
- –20 °C
- Shipping temperature:
- –20 °C
- –20 °C