Order Entry
Export
ContactUsLinkComponent
Anti-dynamin Sheep Polyclonal Antibody
Anti-dynamin Sheep Polyclonal Antibody
Catalog # CAYM10009786-1
Supplier:  Cayman Chemical
Anti-dynamin Sheep Polyclonal Antibody
Catalog # CAYM10009786-1
Supplier:  Cayman Chemical

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Dynamin
  • Antigen symbol:
    DNM
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Sheep
  • Reactivity:
    Rat
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Modification:
    phospho-Ser778
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    phosphopeptide corresponding to amino acid residues surrounding the phospho-Ser778 of rat dynamin
  • Purification:
    purified by affinity chromatography
  • Size:
    1 Ea
  • Pk:
    1

Specifications

About this item

Type: Primary
Antigen: dynamin
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Rat