Order Entry
Export
ContactUsLinkComponent
Anti-NMDA Receptor NR2B subunit Rabbit Polyclonal Antibody
Anti-NMDA Receptor NR2B subunit Rabbit Polyclonal Antibody
Catalog # CAYM10009761-1
Supplier:  Cayman Chemical
Anti-NMDA Receptor NR2B subunit Rabbit Polyclonal Antibody
Catalog # CAYM10009761-1
Supplier:  Cayman Chemical

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    NMDA Receptor NR2B subunit
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Modification:
    phospho-Tyr1472
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    phosphopeptide corresponding to amino acid residues surrounding phospho-Tyr1472 of NMDA Receptor NR2B Subunit
  • Purification:
    purified by affinity chromatography
  • Size:
    1 Ea
  • Pk:
    1

Specifications

About this item

Type: Primary
Antigen: NMDA Receptor NR2B subunit
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat