Anti-RAF-1 Rabbit Polyclonal Antibody
Catalog # CAYM10009505-1
Supplier: Cayman Chemical
Specifications
- Antibody type:Primary
- Antigen name:RAF proto-oncogene serine/threonine-protein kinase
- Antigen symbol:RAF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat
- Western blot:Yes
- Cross adsorption:No
- Modification:phospho-Ser642
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:phosphopeptide corresponding to amino acid residues surrounding phospho-Ser642 of rat raf-1
- Purification:purified by affinity chromatography
- Size:1 Ea
- Pk:1
Specifications
About this item
Type: Primary
Antigen: RAF-1
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat