Order Entry
Export
ContactUsLinkComponent
Anti-TH Rabbit Polyclonal Antibody
Anti-TH Rabbit Polyclonal Antibody
Catalog # CAYM10009413-1
Supplier:  Cayman Chemical
Anti-TH Rabbit Polyclonal Antibody
Catalog # CAYM10009413-1
Supplier:  Cayman Chemical

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    TH
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Modification:
    phospho-Ser31
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    phosphopeptide corresponding to amino acid residues surrounding phospho-Ser31 of rat TH
  • Purification:
    purified by affinity chromatography
  • Size:
    1 Ea
  • Pk:
    1

Specifications

About this item

Type: Primary
Antigen: TH
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat