Order Entry
Export
ContactUsLinkComponent
Anti-CRTH2 Rabbit Polyclonal Antibody
Anti-CRTH2 Rabbit Polyclonal Antibody
Catalog # CAYM10004886-1
Supplier:  Cayman Chemical
Anti-CRTH2 Rabbit Polyclonal Antibody
Catalog # CAYM10004886-1
Supplier:  Cayman Chemical

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    CRTH2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Cross adsorption:
    No
  • Form:
    lyophilized
  • Amino acid number:
    2 to 21
  • Storage buffer:
    TBS, pH 7,4
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    CRTH2/DP2 protein amino acids 2-21
  • Purification:
    purified by affinity chromatography
  • Size:
    1 Ea
  • Pk:
    1

Specifications

About this item

Type: Primary
Antigen: CRTH2
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat