Anti-p53 Rabbit Polyclonal Antibody
CAYM10004807-1
- Antibody type:Primary
- Antigen symbol:P53
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Modification:phospho-Ser392
- Storage buffer:HEPES, pH 7,5, containing 50% glycerol, 0,01% BSA, and 0,15 M sodium chloride
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:amino acids around phospho-Ser392
- Purification:purified by affinity chromatography
- Size:1 Ea
- Pk:1
Type: Primary
Antigen: p53
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human