Anti-Cytokeratin Mouse Monoclonal Antibody [clone: C-11]
Catalog # CAYM10004600-1
Supplier: Cayman Chemical
Specifications
- Antibody type:Primary
- Antigen name:Cytokeratin
- Clonality:Monoclonal
- Clone:C-11
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Storage buffer:PBS, pH 7,4
- Storage temperature:+4 °C
- Shipping temperature:4 °C
- Purification:purified by affinity chromatography
- Size:1 Ea
- Pk:1
Specifications
About this item
Type: Primary
Antigen: Cytokeratin
Clonality: Monoclonal
Clone: C-11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human