Order Entry
Export
ContactUsLinkComponent
Anti-CD45 Mouse Monoclonal Antibody [clone: BRA-55]
Anti-CD45 Mouse Monoclonal Antibody [clone: BRA-55]
Catalog # CAYM10004597-1
Supplier:  Cayman Chemical
Anti-CD45 Mouse Monoclonal Antibody [clone: BRA-55]
Catalog # CAYM10004597-1
Supplier:  Cayman Chemical

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    CD45
  • Clonality:
    Monoclonal
  • Clone:
    BRA-55
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Storage buffer:
    PBS, pH 7,2, containing 0,02% sodium azide
  • Storage temperature:
    +4 °C
  • Shipping temperature:
    4 °C
  • Immunogen:
    human CD45
  • Purification:
    purified by Protein A chromatography
  • Size:
    1 Ea
  • Pk:
    1

Specifications

About this item

Type: Primary
Antigen: CD45
Clonality: Monoclonal
Clone: BRA-55
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human