Anti-Theophylline Rabbit Polyclonal Antibody
Catalog # FITZ20-TR158
Supplier: FITZGERALD INDUSTRIES INTL
Specifications
- Antibody type:Primary
- Antigen name:Theophylline
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Pk:1 mL
Specifications
About this item
Type: Primary
Antigen: Theophylline
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: