Order Entry
Export
ContactUsLinkComponent
Anti-MAP2K1 Rabbit Polyclonal Antibody
Anti-MAP2K1 Rabbit Polyclonal Antibody
  ROCK600-401-D72
undefined
Anti-MAP2K1 Rabbit Polyclonal Antibody
  ROCK600-401-D72

 

  • Antibody type:
    Primary
  • Antigen name:
    Mitogen-activated protein kinase kinase 1
  • Antigen symbol:
    MAP2K1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Antibody fraction
  • Form:
    Liquid
  • Gene ID:
    5604
  • Antigen synonyms:
    MEK1|MAPKK1|CFC3|PRKMK1|MKK1
  • Modification:
    phospho-Thr292
  • Storage buffer:
    0,01 M HEPES, 0,15 M Sodium Chloride, pH 7,50,1 mg/ml Bovine Serum Albumin - IgG and Protease free, 50% (v/v) Glycerol
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Dry Ice
  • Immunogen:
    Anti-MEK1 pT292 Antibody was produced in rabbit by repeated immunizations with synthetic phospho-peptide corresponding to amino acid residues surrounding Thr292 conjugated to KLH.
  • Purification:
    affinity-purified
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Anti-MEK1 pT292 (Rabbit) antibody is suitable for use in Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 45 kDa in size corresponding to MEK 1 Protein phosphorylated at Thr292.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: MAP2K1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat, Human