Anti-FBXO43 Rabbit Polyclonal Antibody
Catalog # ROCK600-401-485
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:F-box protein 43
- Antigen symbol:FBXO43
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Chimpanzee,Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:286151
- Antigen synonyms:FBX43|ERP1|EMI2
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to a region approximately 60 residues downstream of the amino terminal end of human Fbp5B Protein. Human Fbp5B-2
- Purification:affinity-purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
This affinity-purified antibody has been tested for use in ELISA and western blot. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 105 kDa in size corresponding to Fbp5B Protein by western blotting.
Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:500 - 1:3000; Immunoprecipitation: 1:100;
Type: Primary
Antigen: FBXO43
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Chimpanzee