Anti-CHEK2 Rabbit Polyclonal Antibody
Catalog # ROCK600-401-280
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:Checkpoint Kinase 2
- Antigen symbol:CHEK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:11200
- Antigen synonyms:PP1425|LFS2|RAD53|CDS1|hCds1|HuCds1|CHK2
- Modification:phospho-Thr68
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding aa 64-73 of Human CHK2.
- Purification:affinity-purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
This affinity purified antibody has been tested for use in ELISA and by western blot. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 61 kDa in size corresponding to CHK2 by western blotting.
Recommended Dilutions: ELISA: 1:10000 - 1:70,000; Western Blot: 1:200 - 1:2000; Immunoprecipitation: 1:100;
Type: Primary
Antigen: CHEK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human