Anti-CENPQ Rabbit Polyclonal Antibody
ROCK200-401-B48S
- Antibody type:Primary
- Antigen name:Centromere Protein Q
- Antigen symbol:CENPQ
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:55166
- Antigen synonyms:C6orf139|CENP-Q
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:This Protein A purified antibody was prepared from whole rabbit serum produced by repeated immunizations with full-length human CENP-Q recombinant Protein.
- Purification:Protein A purified
- Size:25 µl
- Pk:25 µl
This Protein A purified antibody has been tested for use in ELISA, immunofluorescence microscopy and western blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 26-31 kDa in size.
Recommended Dilutions: ELISA: 1:5000 1:20000; Western Blot: 1:100 - 1:500;
Type: Primary
Antigen: CENPQ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human