Anti-SHANK3 Mouse Monoclonal Antibody (Unconjugated) [clone: S69-46]
Catalog # ROCK200-301-G45
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:SH3 and multiple ankyrin repeat domains 3
- Antigen symbol:SHANK3
- Clonality:Monoclonal
- Clone:S69-46
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:Internal
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:59312
- Antigen synonyms:Prosap2
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,250% (v/v) Glycerol
- Storage temperature:–20 °C
- Concentration:1 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Dry Ice
- Immunogen:SHANK3 Antibody was produced in mice by repeated immunizations raised against a synthetic peptide corresponding to an internal region of rat Shank3.
- Purification:Protein G purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Anti-SHANK3 Antibody is suitable for use in WB, Immunochemistryand IP. Expect a band approximately ~190kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.
Type: Primary
Antigen: SHANK3
Clonality: Monoclonal
Clone: S69-46
Conjugation: Unconjugated
Epitope: Center
Host: Mouse
Isotype: IgG2b
Reactivity: Mouse, Human, Rat