Anti-GRIA2 Mouse Monoclonal Antibody (Unconjugated) [clone: S21-32]
Catalog # ROCK200-301-F34
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:Glutamate receptor 2
- Antigen symbol:GRIA2
- Clonality:Monoclonal
- Clone:S21-32
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:2891
- Antigen synonyms:GLURB|GluA2|GluR-K2|HBGR2|GLUR2
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,250% (v/v) Glycerol
- Storage temperature:–20 °C
- Concentration:1 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Dry Ice
- Immunogen:GluR2 Antibody was produced in mice by repeated immunizations with a fusion Protein corresponding to a cytoplasmic C-terminus region of rat GluR2.
- Purification:Protein G purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Anti-GluR2 Antibody is suitable for use in WB, IHC, IP, and Immuno-gold EM. Expect a band approximately ~90kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.
Type: Primary
Antigen: GRIA2
Clonality: Monoclonal
Clone: S21-32
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat