Anti-ELOVL6 Rabbit Polyclonal Antibody
Catalog # ROCK600-401-C04
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:ELOVL fatty acid elongase 6
- Antigen symbol:ELOVL6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Bovine,Rat,Mouse
- Western blot:Yes
- Epitope:near the N-terminus
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:79071
- Antigen synonyms:FACE|FAE|LCE
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:ELOVL6 Antibody was produced from whole rabbit serum prepared by repeated immunizations with a peptide near the n-terminus of the human ELOVL6.
- Purification:affinity-purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Anti-ELOVL6 Antibody is suitable in ELISA, WB, and IHC. Specific conditions for reactivity should be optimized by the end user. Expect a band at ~29 - 31 kDa in size corresponding to ELOVL6 by Western blotting in the appropriate cell lysate or extract.
Recommended Dilutions: ELISA: 1:10000 - 1:40000; Western Blot: 1.0 - 2.0 µg/ml; Immunochemistry: 5.0 µg/ml;
Type: Primary
Antigen: ELOVL6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine