Anti-CFAP206 Rabbit Polyclonal Antibody
Catalog # ROCK612-401-E42
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:Cilia and flagella associated protein 206
- Antigen symbol:CFAP206
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat
- Western blot:Yes
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid
- Gene ID:69329
- Antigen synonyms:AU024269|1700003M02Rik
- Modification:phospho-Ser416
- Storage buffer:0,01 M HEPES, 0,15 M Sodium Chloride, pH 7,50,1 mg/ml Bovine Serum Albumin - IgG and Protease free, 50% (v/v) Glycerol
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:Anti-Tau pS416 Antibody was produced in rabbit by repeated immunizations with synthetic phospho-peptide corresponding to iamino acid residues surrounding Ser416 of the Tau proetin.
- Purification:affinity-purified
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Anti-Tau pS416 (Rabbit) antibody is suitable for use in Western Blotting. Expect bands or approximately 59, 65, 68 kDa in size corresponding to tau Proteins phosphorylated at Ser416 in the appropriate cell lysate or extract.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: CFAP206
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat