Anti-MAP2K5 Rabbit Polyclonal Antibody
Catalog # ROCK612-401-D74
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:mitogen activated protein kinase kinase 5
- Antigen symbol:MAP2K5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- ChIP:Yes
- Gene ID:29568
- Antigen synonyms:Mek5
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Anti-MEK5 pS311/pT315 (Rabbit) antibody is suitable for use in Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 49 kDa in size corresponding to MEK5 Protein phosphorylated at Ser311/Thr315.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: MAP2K5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat