Anti-CCNL1 Rabbit Polyclonal Antibody
ROCK200-401-881
- Antibody type:Primary
- Antigen name:cyclin L1
- Antigen symbol:CCNL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Antibody fraction
- Form:Lyophilized
- Gene ID:57018
- Antigen synonyms:ANIA6A|PRO1073|BM-001|ania-6a
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:This Protein A purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic polypeptide corresponding to amino acids 314-369 of Human Cyclin L1a Protein.
- Purification:Protein A purified
- Size:500 µg
- Pk:500 µG
This Protein A purified antibody has been tested for use in ELISA, immunochemistry western blot, immunoprecipitation and immunofluores-cence. Specific conditions for reactivity should be optimized by the end user. Expect a band ~ 59 kDa in size.
Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:2000 - 1:5000; Immunochemistry: 10 mg/ml - 20 µg/ml; Immunoprecipitation: 1:100;
Restore with deionized water (or equivalent); 100 µl
Type: Primary
Antigen: CCNL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human