Order Entry
Export
ContactUsLinkComponent
 
Anti-Calprotectin Mouse Monoclonal Antibody
 
Anti-Calprotectin Mouse Monoclonal Antibody

 

  • Catalog No:
  • USBI143119
  • USBI143119-100
  • Antigen symbol:
  • CALPR
  • CALPR
  • Antigen name:
  • Calprotectin
  • Calprotectin
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1
  • IgG1
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Human MRP8 and MRP14 heterocomplex
  • Human MRP8 and MRP14 heterocomplex
  • Form:
  • lyophilized
  • lyophilized
  • Purification:
  • purified by Protein G chromatography
  • purified by Protein G chromatography
  • Cross adsorption:
  • No
  • No
  • Storage buffer:
  • PBS, pH 7,4, 0,09% sodium azide.
  • PBS, pH 7,4, 0,09% sodium azide.
  • Storage temperature:
  • –20 °C
  • –20 °C