Anti-EPO Rabbit Polyclonal Antibody
Catalog # USBI140164
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:EPO
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Amino acid number:82 to 192
- Storage buffer:PBS, pH 7,2, 0,05% sodium azide, 50% glycerol.
- Sequence:MHHHHHHSSGLVPRGSGMKETAAAKFERQHMDSPDLGTDDDDKAMADIGSEFVEEQAIEVWQGLSLLSEAILQAQALLANSSQPPETLQLHIDKAISGLRSLTSLLRVLGAQKELMSPPDTTPPAPLRTLTVDTFCKLFRVYANFLRGKLKLYTGEVCRRGDR
- Storage temperature:–20 °C
- Concentration:200 ug/ml
- Immunogen:EPO (Val82~Arg192); MHHHHHHSSG LVPRGSGMKE TAAAKFERQH MDSPDLGTDD DDKAMADIGS EF- VEEQAIEVW QGLSLLSEAI LQAQALLANS SQPPETLQLH IDKAISGLRS LTSLLRVLGA QKELMSPPDT TPPAPLRTLT VDTFCKLFRV YANFLRGKLK LYTGEVCRRG DR
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: ELISA: 1:100-200; Western Blot: 1:100-400; Immunochemistry: 1:100-500
Type: Primary
Antigen: EPO
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse,