Anti-CD49e Rat Monoclonal Antibody [clone: 5H10-27 (MFR5)]
Catalog # USBIC2405-02F5-50
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:CD49E
- Clonality:Monoclonal
- Clone:5H10-27 (MFR5)
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Cross adsorption:No
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:–20 °C
- Concentration:~0,5 mg/ml
- Immunogen:(C57BL/6 x A/J)F1 mouse mast cell line
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: CD49e
Clonality: Monoclonal
Clone: 5H10-27 (MFR5)
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse,