Anti-Heat Shock Protein 60, P. falciparum Rabbit Polyclonal Antibody
Recommended dilution: Western Blot:1:2000
Type: Primary
Antigen: Heat Shock Protein 60, P. falciparum
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope: full-length
Host: Rabbit
Isotype: IgG
Reactivity:
- Catalog No:
- USBIH1830-70M-25
- USBIH1830-70M-100
- Antigen symbol:
- HSP60
- HSP60
- Antigen name:
- Heat Shock Protein 60
- Heat Shock Protein 60
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Epitope:
- full-length
- full-length
- Immunogen:
- Recombinant full length P. falciparum Hsp60.
- Recombinant full length P. falciparum Hsp60.
- Form:
- liquid
- liquid
- Purification:
- purified by Protein A chromatography
- purified by Protein A chromatography
- Concentration:
- ~1,83 mg/ml
- ~1,83 mg/ml
- Cross adsorption:
- No
- No
- Storage buffer:
- PBS, pH 7,4, 0,09% sodium azide and 50% glycerol.
- PBS, pH 7,4, 0,09% sodium azide and 50% glycerol.
- Storage temperature:
- –20 °C
- –20 °C