Anti-Integrin Rabbit Polyclonal Antibody
Catalog # USBII7661-47-100
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Integrin
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Western blot:Yes
- Cross adsorption:No
- Format:Serum
- Form:lyophilized powder
- Storage buffer:PBS, pH 7,2
- Storage temperature:–20 °C
- Concentration:~1 mg/ml
- Immunogen:Integrin from Geodia cydonium
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Recommended dilution: ELISA: 1:40000; Western Blot: 1:2000-1:4000; Immunoprecipitation: 1:400; Immunochemistry: 1:400
Type: Primary
Antigen: Integrin
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: