Anti-BD5 Rabbit Polyclonal Antibody
USBIB0903-31-100
- Antibody type:Primary
- Antigen symbol:BD5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Cross adsorption:No
- Format:Serum
- Form:lyophilized powder
- Amino acid number:17 to 51
- Storage buffer:PBS, pH 7,2
- Sequence:ESCKLGRGKCRKECLENEKPDGNCRLNFLCCRQRI
- Storage temperature:–20 °C
- Concentration:~1 mg/ml
- Immunogen:Synthetic human β-Defensin 5 (aa 17-51) (ESCKLGRGKCRKECLENEKPDGNCRLNFLCCRQRI)
- Pk:100 µl
Recommended dilution: ELISA:1:2000
Type: Primary
Antigen: BD5
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human,