Anti-Heat Shock Protein 40 Mouse Monoclonal Antibody [clone: 7i2]
Catalog # USBIH1834-15F
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Heat Shock Protein 40
- Antigen symbol:HSP40
- Clonality:Monoclonal
- Clone:7i2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,1% sodium azide.
- Storage temperature:–20 °C
- Concentration:~1 mg/ml
- Immunogen:Recombinant human Hsp40(1-340aa) purified from E. coli
- Purification:purified by Protein G chromatography
- Pk:100 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:1000-1:2000
Type: Primary
Antigen: Heat Shock Protein 40
Clonality: Monoclonal
Clone: 7i2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human,