Order Entry
Export
ContactUsLinkComponent
 
Anti-Tumor Necrosis Factor Receptor 2, Soluble Rabbit Polyclonal Antibody (Biotin)
 
Anti-Tumor Necrosis Factor Receptor 2, Soluble Rabbit Polyclonal Antibody (Biotin)

 

  • Catalog No:
  • USBIT9162-72A-25
  • USBIT9162-72A-50
  • Antigen symbol:
  • TNFR2
  • TNFR2
  • Antigen name:
  • Tumor Necrosis Factor Receptor 2
  • Tumor Necrosis Factor Receptor 2
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Highly pure (> 98%) recombinant human soluble TNF Receptor II
  • Highly pure (> 98%) recombinant human soluble TNF Receptor II
  • Form:
  • lyophilized powder
  • lyophilized powder
  • Purification:
  • purified by immunoaffinity chromatography
  • purified by immunoaffinity chromatography
  • Concentration:
  • ~0,05 mg/ml
  • ~0,05 mg/ml
  • Cross adsorption:
  • No
  • No
  • Storage buffer:
  • PBS
  • PBS
  • Storage temperature:
  • –20 °C
  • –20 °C