Anti-Interleukin 22 Rabbit Polyclonal Antibody (Biotin)
Catalog # USBII8443-18K-25
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 22
- Antigen symbol:IL22
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Form:lyophilized powder
- Storage buffer:PBS
- Storage temperature:–20 °C
- Concentration:~0,05 mg/ml
- Immunogen:Highly pure (> 98%) recombinant murine IL-22
- Purification:purified by immunoaffinity chromatography
- Size:25 μg
- Pk:25 µG
Specifications
About this item
Recommended dilution: ELISA: 0.15-0. 30 ?g/ml
Type: Primary
Antigen: Interleukin 22
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse,