Order Entry
Export
ContactUsLinkComponent
Anti-Interleukin 17 Goat Polyclonal Antibody
Anti-Interleukin 17 Goat Polyclonal Antibody
Catalog # USBII8439-04H-100
Supplier:  US Biological
Anti-Interleukin 17 Goat Polyclonal Antibody
Catalog # USBII8439-04H-100
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Interleukin 17
  • Antigen symbol:
    IL17
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    liquid
  • Storage buffer:
    PBS, 7.2, 40% glycerol.
  • Storage temperature:
    –20 °C
  • Concentration:
    ~1 mg/ml
  • Immunogen:
    Highly pure (> 98%) recombinant human IL-17a.
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    100 μg
  • Pk:
    100 µG

Specifications

About this item

Recommended dilution: ELISA: 0.5 ?g/ml

Type: Primary
Antigen: Interleukin 17
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human,