Anti-Interleukin 15 Rabbit Polyclonal Antibody
Catalog # USBII8437-04F-100
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 15
- Antigen symbol:IL15
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Cross adsorption:No
- Neutralisation:Yes
- Form:lyophilized powder
- Storage buffer:PBS
- Storage temperature:–20 °C
- Immunogen:Highly pure (> 98%) recombinant murine Interleukin-15.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: >0.5 ?g/ml; Western Blot: 0.1-0.2?g/ml; Neutralization: 0. 09-0.14?g/ml
Type: Primary
Antigen: Interleukin 15
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse,