Anti-Galectin-1 Rabbit Polyclonal Antibody
Supplier: US Biological
Certificates
About this item
Recommended dilution: ELISA: 0.5-2?g/ml; Western Blot: 0.1-0.2?g/ml
Type: Primary
Antigen: Galectin-1
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,
Specifications
- Catalog No:
- USBIG1044-47-100
- USBIG1044-47-50
- Antigen symbol:
- GAL1
- GAL1
- Antigen name:
- Galectin 1
- Galectin 1
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Highly pure (>98%) recombinant human Galectin-1.
- Highly pure (>98%) recombinant human Galectin-1.
- Form:
- lyophilized powder
- lyophilized powder
- Purification:
- purified by immunoaffinity chromatography
- purified by immunoaffinity chromatography
- Concentration:
- ~1 mg/ml
- ~1 mg/ml
- Cross adsorption:
- No
- No
- Storage buffer:
- PBS, pH 7,2.
- PBS, pH 7,2.
- Storage temperature:
- –20 °C
- –20 °C