Anti-Bone Morphogenic Protein 14 Rabbit Polyclonal Antibody
USBIB2553-50D-100
- Antibody type:Primary
- Antigen name:Bone Morphogenic Protein 14
- Antigen symbol:BMP14
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Serum
- Form:liquid
- Storage buffer:PBS, 40% glycerol.
- Storage temperature:–20 °C
- Immunogen:Recombinant purified human BMP-14/CDMP-1 protein.
- Size:100 μl
- Pk:100 µl
Recommended dilution: ELISA: 1:10000-1:50000; Western Blot: 1:1000-1:5000
Type: Primary
Antigen: Bone Morphogenic Protein 14
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,