Anti-MAP Kinase p44, p42, Mouse Monoclonal Antibody (Biotin) [clone: 6D69]
Catalog # USBIM2363-01A-100
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:MAP Kinase p44, p42,
- Clonality:Monoclonal
- Clone:6D69
- Conjugation:Biotin
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human,Dog,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Modification:phospho-Thr202, phospho-Tyr204
- Storage buffer:PBS, 0,09% sodium azide, PEG, Sucrose and 40% glycerol
- Storage temperature:–20 °C
- Immunogen:Synthetic peptide corresponding to the FLpTEpYVA motif of phosphorylated ERK1/2 (MAPK2/3). Conjugated to KLH.
- Purification:purified by subsequent thiophillic adsorption and size exclusion chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: 0. 05 ?g/ml; Western Blot: 0.5 ?g/ml
Type: Primary
Antigen: MAP Kinase p44, p42,
Clonality: Monoclonal
Clone: 6D69
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Mouse, Rat,