Order Entry
Export
ContactUsLinkComponent
Anti-Heat Shock Protein 27, Mouse Monoclonal Antibody [clone: 6D435]
Anti-Heat Shock Protein 27, Mouse Monoclonal Antibody [clone: 6D435]
Catalog # USBIH1830-52K
Supplier:  US Biological
Anti-Heat Shock Protein 27, Mouse Monoclonal Antibody [clone: 6D435]
Catalog # USBIH1830-52K
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Heat shock protein 27
  • Antigen symbol:
    HSP27
  • Clonality:
    Monoclonal
  • Clone:
    6D435
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Dog,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Amino acid number:
    79 to 85
  • Modification:
    phospho-Ser82
  • Immunogen:
    Synthetic peptide corresponding to aa 79-85 (R79QLpSSGV85) of HSP27 phosphorylated at Ser82.
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: Heat Shock Protein 27,
Clonality: Monoclonal
Clone: 6D435
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Mouse, Rat,