Anti-Catenin, beta, Exon 14 Mouse Monoclonal Antibody [clone: 6D209]
Catalog # USBIC2069-52T
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Catenin, beta, Exon 14
- Clonality:Monoclonal
- Clone:6D209
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Immunogen:Recombinant β-catenin.
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: Catenin, beta, Exon 14
Clonality: Monoclonal
Clone: 6D209
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat,