Anti-Interleukin 13 Rat Monoclonal Antibody [clone: 5K214]
USBII8435-05A-250
- Antibody type:Primary
- Antigen name:Interleukin 13
- Antigen symbol:IL13
- Clonality:Monoclonal
- Clone:5K214
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rat
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Storage buffer:borate buffered saline, pH 8,2
- Storage temperature:–20 °C
- Concentration:0,5 mg/ml
- Immunogen:Recombinant corresponding to human Interleukin 13 expressed in COS cells.
- Purification:purified
- Size:250 μg
- Pk:250 µG
Type: Primary
Antigen: Interleukin 13
Clonality: Monoclonal
Clone: 5K214
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1 kappa
Reactivity: Human,